Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAE2
Entrez Gene ID
820649
Gene Name
20S proteasome alpha subunit E2
Gene Aliases
20S PROTEASOME SUBUNIT PAE2, 20S proteasome alpha subunit E2, AT3G14290
Chromosome Location
Chr.3: 4764024 - 4766609 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112287.4 NP_188046.1 7 - 8 686 63
GenBank mRNA AF043525.1 - 6 - 7 491 63
AF370245.1 - 7 - 8 552 63
AY062970.1 - 5 - 6 449 63

View Full Product Details