Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATPMEPCRC
- Entrez Gene ID
-
820650
- Gene Name
- pectinesterase family protein
- Gene Aliases
-
A. THALIANA PECTIN METHYLESTERASE 26, AT3G14300, ATPME26, PECTIN METHYLESTERASE 26, PME26
- Chromosome Location
-
Chr.3: 4766905 - 4769936 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112288.2 |
NP_188047.1 |
1 - 2 |
2262 |
130 |
View Full Product Details