Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP25
Entrez Gene ID
820662
Gene Name
ubiquitin-specific protease 25
Gene Aliases
AT3G14400, UBIQUITIN-SPECIFIC PROTEASE 25, ubiquitin-specific protease 25
Chromosome Location
Chr.3: 4811486 - 4815515 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112299.4 NP_566486.1 6 - 7 2050 93
GenBank mRNA AF302673.1 - 6 - 7 1745 93
BT021128.1 - 6 - 7 1766 93

View Full Product Details