Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP72A8
- Entrez Gene ID
-
820690
- Gene Name
- cytochrome P450, family 72, subfamily A, polypeptide 8
- Gene Aliases
-
''cytochrome P450, AT3G14620, cytochrome P450, family 72, polypeptide 8, polypeptide 8'', subfamily A
- Chromosome Location
-
Chr.3: 4914789 - 4917176 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112323.5 |
NP_188080.1 |
4 - 5 |
1315 |
96 |
GenBank mRNA |
AY052208.1 |
- |
4 - 5 |
1179 |
96 |
View Full Product Details