Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHM3
Entrez Gene ID
820707
Gene Name
rhamnose biosynthesis 3
Gene Aliases
ARABIDOPSIS THALIANA RHAMNOSE BIOSYNTHESIS 3, AT3G14790, ATRHM3, rhamnose biosynthesis 3
Chromosome Location
Chr.3: 4964105 - 4967075 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338136.1 NP_001326092.1 2 - 3 2061 121
NM_112340.4 NP_188097.1 2 - 3 1751 121
GenBank mRNA AY078958.1 - 2 - 3 1704 121
AY142060.1 - 1 - 2 1551 121

View Full Product Details