Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPC3
Entrez Gene ID
820723
Gene Name
phosphoenolpyruvate carboxylase 3
Gene Aliases
AT3G14940, ATPPC3, phosphoenolpyruvate carboxylase 3
Chromosome Location
Chr.3: 5025184 - 5029886 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338141.1 NP_001326778.1 5 - 6 1180 85
NM_112356.4 NP_188112.1 6 - 7 1206 85
GenBank mRNA AF071788.1 - 6 - 7 1060 85

View Full Product Details