Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTL2
Entrez Gene ID
820724
Gene Name
tetratricopetide-repeat thioredoxin-like 2
Gene Aliases
AT3G14950, tetratricopetide-repeat thioredoxin-like 2
Chromosome Location
Chr.3: 5029987 - 5033102 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338142.1 NP_001319556.1 6 - 7 2244 75

View Full Product Details