Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ERF4
- Entrez Gene ID
-
820752
- Gene Name
- ethylene responsive element binding factor 4
- Gene Aliases
-
AT3G15210, ATERF-4, ATERF4, ETHYLENE RESPONSIVE ELEMENT BINDING FACTOR 4, RAP2.5, RELATED TO AP2 5, ethylene responsive element binding factor 4
- Chromosome Location
-
Chr.3: 5121303 - 5122562 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112384.2 |
NP_188139.1 |
1 - 1 |
1075 |
151 |
View Full Product Details