Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPI2
Entrez Gene ID
820765
Gene Name
proton pump interactor 2
Gene Aliases
AT3G15340, proton pump interactor 2
Chromosome Location
Chr.3: 5159698 - 5162945 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125161.1 NP_001118633.1 3 - 4 987 93
NM_001338153.1 NP_001326301.1 4 - 5 1193 93
NM_112397.4 NP_188152.3 5 - 6 1165 93

View Full Product Details