Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY18
Entrez Gene ID
829308
Gene Name
WRKY DNA-binding protein 18
Gene Aliases
ARABIDOPSIS THALIANA WRKY DNA-BINDING PROTEIN 18, AT4G31800, ATWRKY18, F11C18.16, WRKY DNA-binding protein 18
Chromosome Location
Chr.4: 15382777 - 15385100 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342116.1 NP_001329847.1 1 - 2 167 93
NM_119329.4 NP_567882.1 1 - 2 587 93
GenBank mRNA AF428421.1 - 1 - 2 161 93
AK227401.1 - 1 - 2 161 93
AY125557.1 - 1 - 2 68 93

View Full Product Details