Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GPX7
- Entrez Gene ID
-
829316
- Gene Name
- glutathione peroxidase 7
- Gene Aliases
-
AT4G31870, ATGPX7, F11C18.70, F11C18_70, GLUTATHIONE PEROXIDASE 7, glutathione peroxidase 7
- Chromosome Location
-
Chr.4: 15410078 - 15412486 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119337.3 |
NP_194915.2 |
3 - 4 |
509 |
78 |
View Full Product Details