Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RR10
Entrez Gene ID
829322
Gene Name
response regulator 10
Gene Aliases
ARR10, AT4G31920, F11C18.3, response regulator 10
Chromosome Location
Chr.4: 15444053 - 15447042 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119343.4 NP_194920.1 3 - 4 891 112
GenBank mRNA AJ005195.1 - 3 - 4 701 112
AK228149.1 - 3 - 4 772 112
BT003158.1 - 3 - 4 615 112

View Full Product Details