Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASY2
Entrez Gene ID
829353
Gene Name
DNA-binding HORMA family protein
Gene Aliases
AT4G32200, F10M6.160, F10M6_160
Chromosome Location
Chr.4: 15548840 - 15555105 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342129.1 NP_001329706.1 2 - 3 134 72
NM_119372.3 NP_194947.2 2 - 3 134 72

View Full Product Details