Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AME3
Entrez Gene ID
829401
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT4G32660, F4D11.140, F4D11_140
Chromosome Location
Chr.4: 15756018 - 15759314 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036697.1 NP_001031774.1 2 - 3 232 100
NM_001342166.1 NP_001328058.1 1 - 2 468 100
NM_119418.3 NP_194992.1 1 - 2 468 100
NM_202937.1 NP_974666.1 1 - 2 358 100
GenBank mRNA BT029776.1 - 1 - 2 90 100
D45355.1 - 1 - 2 149 100

View Full Product Details