Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- VTE1
- Entrez Gene ID
-
829413
- Gene Name
- tocopherol cyclase, chloroplast / vitamin E deficient 1 (VTE1) / sucrose export defective 1 (SXD1)
- Gene Aliases
-
AT4G32770, ATSDX1, F4D11.30, F4D11_30, SUCROSE EXPORT DEFECTIVE 1, VITAMIN E DEFICIENT 1
- Chromosome Location
-
Chr.4: 15804784 - 15808084 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119430.5 |
NP_567906.1 |
5 - 6 |
1012 |
67 |
GenBank mRNA |
AF302188.1 |
- |
5 - 6 |
815 |
67 |
View Full Product Details