Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PME44
Entrez Gene ID
829458
Gene Name
pectin methylesterase 44
Gene Aliases
A. THALIANA PECTIN METHYLESTERASE 44, AT4G33220, ATPME44, F4I10.150, F4I10_150, pectin methylesterase 44
Chromosome Location
Chr.4: 16022269 - 16026428 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342204.1 NP_001328318.1 2 - 3 2011 83
NM_119476.7 NP_567917.4 2 - 3 653 83
GenBank mRNA AF378892.1 - 2 - 3 482 83
BT029770.1 - 1 - 2 53 83

View Full Product Details