Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DHS2
Entrez Gene ID
829489
Gene Name
3-deoxy-d-arabino-heptulosonate 7-phosphate synthase
Gene Aliases
3-DEOXY-D-ARABINO-HEPTULOSONATE 7-PHOSPHATE SYNTHASE, 3-DEOXY-D-ARABINO-HEPTULOSONATE-7-PHOSPHATE 2, 3-deoxy-d-arabino-heptulosonate 7-phosphate synthase, AT4G33510, AtDAHP2, DAHP2, F17M5.270, F17M5_270
Chromosome Location
Chr.4: 16116371 - 16118825 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342217.1 NP_001320125.1 2 - 3 785 82
NM_119505.3 NP_195077.1 2 - 3 861 82
GenBank mRNA AF361798.1 - 2 - 3 785 82
AY124858.1 - 2 - 3 736 82

View Full Product Details