Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCA5
Entrez Gene ID
829498
Gene Name
beta carbonic anhydrase 5
Gene Aliases
A. THALIANA BETA CARBONIC ANHYDRASE 5, AT4G33580, ATBCA5, T16L1.70, T16L1_70, beta carbonic anhydrase 5
Chromosome Location
Chr.4: 16138251 - 16141624 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036707.1 NP_001031784.1 7 - 8 706 69
NM_001342222.1 NP_001328574.1 6 - 7 720 69
NM_119514.4 NP_567928.1 7 - 8 812 69
GenBank mRNA AK221870.1 - 7 - 8 704 69
AY034930.1 - 6 - 7 567 69
AY088420.1 - 6 - 7 604 69
AY122986.1 - 6 - 7 512 69

View Full Product Details