Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRP3A
Entrez Gene ID
829506
Gene Name
dynamin-related protein 3A
Gene Aliases
ABERRANT PEROXISOME MORPHOLOGY 1, ADL2, ADL2A, APEM1, ARABIDOPSIS DYNAMIN-LIKE 2, AT4G33650, NOXY15, T16L1.140, T16L1_140, dynamin-related protein 3A, non responding to oxylipins 15
Chromosome Location
Chr.4: 16160824 - 16166994 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203977.2 NP_001190906.1 19 - 20 2499 66
NM_119521.5 NP_567931.1 19 - 20 2482 66
GenBank mRNA AB072373.1 - 19 - 20 2337 66

View Full Product Details