Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OST1
Entrez Gene ID
829541
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT4G33950, ATOST1, F17I5.140, F17I5_140, OPEN STOMATA 1, P44, SNF1-RELATED PROTEIN KINASE 2.6, SNRK2-6, SNRK2.6, SRK2E, SUCROSE NONFERMENTING 1-RELATED PROTEIN KINASE 2-6
Chromosome Location
Chr.4: 16271745 - 16274825 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342244.1 NP_001320129.1 7 - 8 1137 132
NM_119556.3 NP_567945.1 7 - 8 1309 132
GenBank mRNA AY054624.1 - 7 - 8 735 132
AY081538.1 - 7 - 8 690 132

View Full Product Details