Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABF3
Entrez Gene ID
829547
Gene Name
abscisic acid responsive elements-binding factor 3
Gene Aliases
AT4G34000, AtABF3, DC3 PROMOTER-BINDING FACTOR 5, DPBF5, F17I5.200, abscisic acid responsive elements-binding factor 3
Chromosome Location
Chr.4: 16295334 - 16298285 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342246.1 NP_001320130.1 3 - 4 1927 96
NM_119562.5 NP_567949.1 4 - 5 1390 96
NM_179159.4 NP_849490.2 4 - 5 1595 96
GenBank mRNA AF093546.1 - 3 - 4 1290 96
AK175851.1 - 4 - 5 1341 96
AY054605.1 - 4 - 5 1326 96
AY081467.1 - 3 - 4 1253 96

View Full Product Details