Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCCB
Entrez Gene ID
829549
Gene Name
3-methylcrotonyl-CoA carboxylase
Gene Aliases
3-METHYLCROTONYL-COA CARBOXYLASE, 3-methylcrotonyl-CoA carboxylase, AT4G34030, F28A23.210, F28A23_210
Chromosome Location
Chr.4: 16301027 - 16304099 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119564.5 NP_567950.1 4 - 5 1153 60
GenBank mRNA AF059510.1 - 4 - 5 960 60
BT000384.1 - 4 - 5 882 60

View Full Product Details