Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARI1
Entrez Gene ID
829587
Gene Name
RING/U-box superfamily protein
Gene Aliases
ARABIDOPSIS ARIADNE 1, ARIADNE 1, AT4G34370, ATARI1, F10M10.140, F10M10_140
Chromosome Location
Chr.4: 16434280 - 16437415 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342281.1 NP_001320134.1 2 - 3 428 83
NM_119602.2 NP_567966.1 2 - 3 406 83
GenBank mRNA AY050864.1 - 2 - 3 375 83
AY117264.1 - 2 - 3 161 83

View Full Product Details