Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RRTF1
- Entrez Gene ID
-
829591
- Gene Name
- redox responsive transcription factor 1
- Gene Aliases
-
AT4G34410, redox responsive transcription factor 1
- Chromosome Location
-
Chr.4: 16451876 - 16453260 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119606.2 |
NP_195167.1 |
1 - 1 |
1242 |
157 |
- Gene Symbol
- AT4G34412
- Entrez Gene ID
-
829592
- Gene Name
- EKC/KEOPS complex subunit tprkb-like protein
- Gene Aliases
-
AT4G34412
- Chromosome Location
-
Chr.4: 16452960 - 16454633 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_148397.2 |
NP_680763.2 |
6 - 6 |
619 |
157 |
GenBank mRNA |
BT010697.1 |
- |
6 - 6 |
565 |
157 |
View Full Product Details