Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAP1
Entrez Gene ID
829600
Gene Name
cyclase associated protein 1
Gene Aliases
AT4G34490, ATCAP1, CAP 1, T4L20.70, T4L20_70, cyclase associated protein 1
Chromosome Location
Chr.4: 16484470 - 16487574 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342287.1 NP_001329504.1 4 - 5 1054 95
NM_119614.6 NP_195175.1 4 - 5 1013 95
GenBank mRNA AB014759.1 - 4 - 5 890 95
BT029161.1 - 4 - 5 835 95

View Full Product Details