Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BLH6
Entrez Gene ID
829613
Gene Name
BEL1-like homeodomain 6
Gene Aliases
AT4G34610, BEL1-like homeodomain 6, T4L20.190, T4L20_190
Chromosome Location
Chr.4: 16530346 - 16533362 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125644.1 NP_001119116.1 2 - 3 912 88
NM_001342295.1 NP_001328814.1 1 - 2 1509 88
NM_001342296.1 NP_001328815.1 2 - 3 984 88
NM_119627.4 NP_195187.1 2 - 3 1297 88
GenBank mRNA BT012291.1 - 1 - 2 645 88

View Full Product Details