Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SQS2
Entrez Gene ID
829617
Gene Name
squalene synthase 2
Gene Aliases
AT4G34650, T4L20.230, T4L20_230, squalene synthase 2
Chromosome Location
Chr.4: 16542242 - 16545194 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342298.1 NP_001328690.1 1 - 2 121 88
NM_119631.3 NP_195191.2 5 - 6 602 88

View Full Product Details