Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPSF73-I
Entrez Gene ID
842393
Gene Name
cleavage and polyadenylation specificity factor 73-I
Gene Aliases
AT1G61010, T7P1.15, T7P1_15, cleavage and polyadenylation specificity factor 73-I
Chromosome Location
Chr.1: 22474267 - 22477765 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333924.1 NP_001319278.1 5 - 6 2517 70
NM_179504.2 NP_849835.1 6 - 7 2257 70
GenBank mRNA AK316692.1 - 6 - 7 2194 70

View Full Product Details