Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLP1
Entrez Gene ID
844025
Gene Name
tubby like protein 1
Gene Aliases
AT1G76900, AtTLP1, F7O12.7, tubby like protein 1
Chromosome Location
Chr.1: 28881539 - 28885023 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334747.1 NP_001319391.1 3 - 4 1717 97
NM_106341.2 NP_177816.1 4 - 5 650 97
NM_179563.3 NP_849894.1 4 - 5 1062 97
GenBank mRNA AF487267.1 - 3 - 4 577 97
AY139758.1 - 4 - 5 748 97
BT003039.1 - 3 - 4 577 97

View Full Product Details