Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACR3
Entrez Gene ID
844035
Gene Name
ACT domain repeat 3
Gene Aliases
ACT domain repeat 3, AT1G76990, F22K20.9, F22K20_9
Chromosome Location
Chr.1: 28932119 - 28936371 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036212.1 NP_001031289.1 6 - 7 1291 64
NM_001124136.2 NP_001117608.1 6 - 7 1954 64
NM_106350.3 NP_565146.1 6 - 7 1334 64
NM_179565.2 NP_849896.1 6 - 7 1445 64
NM_179566.2 NP_849897.1 5 - 6 1541 64
GenBank mRNA AF360322.1 - 6 - 7 1265 64
AF528059.1 - 6 - 7 1214 64
AK317369.1 - 5 - 6 1524 64
AY113891.1 - 5 - 6 1022 64

View Full Product Details