Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIE1
Entrez Gene ID
814703
Gene Name
RING-finger protein for embryogenesi
Gene Aliases
AT2G01735, RING-finger protein for embryogenesis
Chromosome Location
Chr.2: 324365 - 326094 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335075.1 NP_001325257.1 3 - 4 1128 68
NM_179593.4 NP_849924.1 4 - 5 1048 68
GenBank mRNA AY168924.1 - 4 - 5 941 68

View Full Product Details