The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001335657.1 | NP_001323708.1 | 1 - 2 | 504 | 102 |
NM_179663.3 | NP_849994.1 | 1 - 2 | 504 | 102 | |
GenBank mRNA | AJ487669.1 | - | 1 - 2 | 504 | 102 |