Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLASP
Entrez Gene ID
816539
Gene Name
CLIP-associated protein
Gene Aliases
AT2G20190, ATCLASP, CLIP-ASSOCIATED PROTEIN, CLIP-associated protein, T2G17.1, T2G17_1
Chromosome Location
Chr.2: 8711579 - 8719115 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179666.5 NP_849997.2 3 - 4 847 114
GenBank mRNA AY091025.1 - 3 - 4 740 114

View Full Product Details