Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALE2
Entrez Gene ID
816549
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT2G20300, Abnormal Leaf Shape 2, F11A3.15, F11A3_15
Chromosome Location
Chr.2: 8755852 - 8760298 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179667.3 NP_849998.1 6 - 7 1760 66
GenBank mRNA AK226880.1 - 6 - 7 1682 66
AY091071.1 - 6 - 7 1681 66
AY133858.1 - 6 - 7 1307 66

View Full Product Details