Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RKP
Entrez Gene ID
816737
Gene Name
related to KPC1
Gene Aliases
AT2G22010, AT2G22020, F7D8.33, F7D8_33, related to KPC1
Chromosome Location
Chr.2: 9363109 - 9369461 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202644.1 NP_001189573.1 6 - 7 3126 137
NM_179689.3 NP_850020.1 6 - 7 3113 137

View Full Product Details