Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAI1
Entrez Gene ID
816807
Gene Name
basic helix-loop-helix (bHLH) DNA-binding superfamily protein
Gene Aliases
AT2G22770, T30L20.3, T30L20_3
Chromosome Location
Chr.2: 9684562 - 9686613 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179700.3 NP_850031.2 2 - 3 1192 145
GenBank mRNA AF488564.1 - 2 - 3 896 145

View Full Product Details