Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OVA4
Entrez Gene ID
817126
Gene Name
Nucleotidylyl transferase superfamily protein
Gene Aliases
AT2G25840, F17H15.13, F17H15_13, ovule abortion 4
Chromosome Location
Chr.2: 11021692 - 11025451 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179739.4 NP_850070.1 12 - 13 1319 81
NM_201804.3 NP_973533.1 12 - 13 1175 81
NM_201805.1 NP_973534.1 12 - 13 1070 81
GenBank mRNA AY081299.1 - 12 - 13 1106 81
BT001221.1 - 12 - 13 1087 81

View Full Product Details