Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CAM5
- Entrez Gene ID
-
817245
- Gene Name
- calmodulin 5
- Gene Aliases
-
1357, ACAM-2, AT2G27030, CAM2, T20P8.8, calmodulin 2, calmodulin 5
- Chromosome Location
-
Chr.2: 11531967 - 11534358 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179766.4 |
NP_850097.1 |
2 - 3 |
551 |
101 |
View Full Product Details