Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRS3
Entrez Gene ID
817253
Gene Name
FAR1-related sequence 3
Gene Aliases
AT2G27110, FAR1-related sequence 3, T20P8.16, T20P8_16
Chromosome Location
Chr.2: 11576668 - 11580817 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036351.2 NP_001031428.1 6 - 7 2481 92
NM_128269.4 NP_565636.1 5 - 6 2778 92
NM_179767.2 NP_850098.1 4 - 5 2925 92
GenBank mRNA AK317350.1 - 6 - 7 2424 92
AY057491.1 - 5 - 6 2716 92
BT002690.1 - 4 - 5 2513 92

View Full Product Details