Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGO5
Entrez Gene ID
817334
Gene Name
Argonaute family protein
Gene Aliases
ARGONAUTE 5, AT2G27880, AtAGO5, T1E2.20, T1E2_20
Chromosome Location
Chr.2: 11871378 - 11876989 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179779.3 NP_850110.1 17 - 18 2725 136

View Full Product Details