Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ftsh3
Entrez Gene ID
817456
Gene Name
FTSH protease 3
Gene Aliases
AT2G29080, FTSH protease 3, T9I4.16, T9I4_16
Chromosome Location
Chr.2: 12489627 - 12493329 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179798.2 NP_850129.1 4 - 5 805 134
GenBank mRNA BT002743.1 - 4 - 5 475 134

View Full Product Details