The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001336220.1 | NP_001325344.1 | 3 - 4 | 962 | 76 |
NM_128519.6 | NP_180526.1 | 4 - 5 | 915 | 76 | |
NM_179805.2 | NP_850136.1 | 4 - 5 | 930 | 76 | |
NM_201832.1 | NP_973561.1 | 4 - 5 | 728 | 76 | |
GenBank mRNA | BT001983.1 | - | 4 - 5 | 889 | 76 |
BT008489.1 | - | 4 - 5 | 841 | 76 |