Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HVT1
- Entrez Gene ID
-
817631
- Gene Name
- helicase in vascular tissue and tapetum
- Gene Aliases
-
AT2G30800, ATVT-1, helicase in vascular tissue and tapetum
- Chromosome Location
-
Chr.2: 13120292 - 13126813 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179823.4 |
NP_850154.2 |
2 - 3 |
386 |
150 |
View Full Product Details