Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATX1
Entrez Gene ID
817721
Gene Name
homologue of trithorax
Gene Aliases
AT2G31650, SDG27, SET DOMAIN PROTEIN 27, T9H9.17, T9H9_17, homologue of trithorax
Chromosome Location
Chr.2: 13455272 - 13462431 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179839.3 NP_850170.1 6 - 7 1413 83

View Full Product Details