Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDE5
Entrez Gene ID
820778
Gene Name
silencing defective 5
Gene Aliases
AT3G15390, silencing defective 5
Chromosome Location
Chr.3: 5196522 - 5200394 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202971.2 NP_001189900.1 9 - 10 947 97
NM_001338166.1 NP_001327652.1 10 - 11 1556 97
NM_112407.5 NP_188158.4 11 - 12 1601 97

View Full Product Details