Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATA20
Entrez Gene ID
820779
Gene Name
anther 20
Gene Aliases
AT3G15400, anther 20
Chromosome Location
Chr.3: 5201622 - 5203197 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338169.1 NP_001319559.1 2 - 2 1441 90
NM_112408.3 NP_188159.1 1 - 1 1517 90

View Full Product Details