The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035626.2 | NP_001030703.1 | 3 - 4 | 605 | 82 |
NM_112432.2 | NP_566520.1 | 3 - 4 | 780 | 82 |