Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRX4
Entrez Gene ID
820809
Gene Name
glutaredoxin 4
Gene Aliases
A. THALIANA GLUTAREDOXIN 4, AT3G15660, ATGRX4, glutaredoxin 4
Chromosome Location
Chr.3: 5307694 - 5309624 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035627.3 NP_001030704.1 6 - 7 620 64
NM_112436.3 NP_566522.1 5 - 6 697 64
GenBank mRNA AK227628.1 - 5 - 6 566 64
AK317312.1 - 6 - 7 553 64
BT006396.1 - 5 - 6 456 64

View Full Product Details