Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PnsB3
Entrez Gene ID
820871
Gene Name
NDH-dependent cyclic electron flow 1
Gene Aliases
AT3G16250, NDF4, NDH-dependent cyclic electron flow 1, Photosynthetic NDH subcomplex B 3
Chromosome Location
Chr.3: 5506931 - 5508414 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112496.3 NP_188246.1 4 - 5 550 74
GenBank mRNA AY042867.1 - 4 - 5 551 74
BT006260.1 - 4 - 5 456 74

View Full Product Details