Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCG29
Entrez Gene ID
820881
Gene Name
pleiotropic drug resistance 1
Gene Aliases
AT3G16340, ATP-binding cassette G29, ATPDR1, AtABCG29, PDR1, PLEIOTROPIC DRUG RESISTANCE 1, pleiotropic drug resistance 1
Chromosome Location
Chr.3: 5539385 - 5546487 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202982.2 NP_001189911.1 1 - 2 577 84
NM_112505.4 NP_566543.1 1 - 2 823 84

View Full Product Details